Thermo Scientific Chemicals

2-Ethylthiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13372.22
100 g, Each
Thermo Scientific Chemicals

2-Ethylthiophene, 99%, Thermo Scientific Chemicals

Catalog number: A13372.22
100 g, Each
Quantity
Catalog number: A13372.22
also known as A13372-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-55-9
IUPAC Name
2-ethylthiophene
Molecular Formula
C6H8S
InChI Key
JCCCMAAJYSNBPR-UHFFFAOYSA-N
SMILES
CCC1=CC=CS1
Identification (FTIR)
Conforms
Refractive Index
1.5115-1.5145 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid

Description

It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text