Thermo Scientific Chemicals

6-Methylheptane-2,4-dione, 98+%, Thermo Scientific Chemicals

Catalog number: A13373.09
10 g, Each
Thermo Scientific Chemicals

6-Methylheptane-2,4-dione, 98+%, Thermo Scientific Chemicals

Catalog number: A13373.09
10 g, Each
Quantity
Catalog number: A13373.09
also known as A13373-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3002-23-1
IUPAC Name
6-methylheptane-2,4-dione
Molecular Formula
C8H14O2
InChI Key
IGMOYJSFRIASIE-UHFFFAOYSA-N
SMILES
CC(C)CC(=O)CC(C)=O
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Refractive Index
1.4550-1.4590 @ 20?C
Identification (FTIR)
Conforms

Description

6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text