Thermo Scientific Chemicals

4-Fluorobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog number: A13377.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog number: A13377.14
25 g, Each
Quantity
Catalog number: A13377.14
also known as A13377-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-44-8
IUPAC Name
1-fluoro-4-(trifluoromethyl)benzene
Molecular Formula
C7H4F4
InChI Key
UNNNAIWPDLRVRN-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)C(F)(F)F
Assay (GC)
≥98.0%
Refractive Index
1.3995-1.4035 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

4-Fluorobenzotrifluoride was used in the synthesis of 2,2?-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2?-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text