Thermo Scientific Chemicals

1,3-Dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13380.22
100 g, Each
Thermo Scientific Chemicals

1,3-Dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13380.22
100 g, Each
Quantity
Catalog number: A13380.22
also known as A13380-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151-10-0
IUPAC Name
1,3-dimethoxybenzene
Molecular Formula
C8H10O2
InChI Key
DPZNOMCNRMUKPS-UHFFFAOYSA-N
SMILES
COC1=CC(OC)=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Assay from Suppliers CofA
≥97.5%
Form
Liquid

Description

1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dimethoxybenzene is used for the preparation of oxathiane spiroketal donors. It is also used for the formation of pi- and O-ylidic complexes with dichlorocarbene. Further, it acts as a flavoring agent.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text