Thermo Scientific Chemicals

5-Bromothiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13381.06
5 g, Each
Thermo Scientific Chemicals

5-Bromothiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13381.06
5 g, Each
Quantity
Catalog number: A13381.06
also known as A13381-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4701-17-1
IUPAC Name
5-bromothiophene-2-carbaldehyde
Molecular Formula
C5H3BrOS
InChI Key
GFBVUFQNHLUCPX-UHFFFAOYSA-N
SMILES
BrC1=CC=C(S1)C=O
Appearance (Color)
Clear, pale yellow to yellow to orange or brown to dark brown
Assay (GC)
>96.0%
Free acid (titration)
<1.0%
Refractive Index
1.6365-1.6415 @ 20?C
Form
Liquid

Description

5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

Solubility
Soluble in chloroform and methanol.

Notes
Air Sensitive, avoid contact with air/oxygen. Store in cool, dry place in tightly closed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text