4,5-Dibromothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals
4,5-Dibromothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,5-Dibromothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A13384.06
also known as A13384-06
Price (USD)/ Each
169.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
169.00
Add to cart
4,5-Dibromothiophene-2-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberA13384.06
Price (USD)/ Each
169.00
-
Add to cart
Chemical Identifiers
CAS81606-31-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormPowder
Appearance (Color)White to pale cream
Melting Point (clear melt)72.0-81.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only