Thermo Scientific Chemicals

1-Bromo-3-methyl-2-butene, 90+%, stab. with silver, Thermo Scientific Chemicals

Catalog number: A13388.09
10 g, Each
Thermo Scientific Chemicals

1-Bromo-3-methyl-2-butene, 90+%, stab. with silver, Thermo Scientific Chemicals

Catalog number: A13388.09
10 g, Each
Quantity
Catalog number: A13388.09
also known as A13388-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
870-63-3
IUPAC Name
1-bromo-3-methylbut-2-ene
Molecular Formula
C5H9Br
InChI Key
LOYZVRIHVZEDMW-UHFFFAOYSA-N
SMILES
CC(C)=CCBr
Assay (GC)
> 90.0%
Appearance (Color)
Colourless to yellow
Refractive Index
1.4855-1.4935
Form
liquid

Description

1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.

Solubility
Immiscible with water

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text