Thermo Scientific Chemicals

Diethanolamine, 99%, Thermo Scientific Chemicals

Catalog number: A13389.0E
2500 g, Each
Thermo Scientific Chemicals

Diethanolamine, 99%, Thermo Scientific Chemicals

Catalog number: A13389.0E
2500 g, Each
Quantity
Catalog number: A13389.0E
also known as A13389-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-42-2
IUPAC Name
2-[(2-hydroxyethyl)amino]ethan-1-ol
Molecular Formula
C4H11NO2
InChI Key
ZBCBWPMODOFKDW-UHFFFAOYSA-N
SMILES
OCCNCCO
Appearance (Color)
Clear colorless
Form
Liquid or viscous liquid
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Identification (FTIR)
Conforms (non-U.S. specification)

Description

Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethanolamine is used in the preparation of morpholine and diethanolamides, which is an active ingredient in cosmetics and shampoos. It acts as a surfactant and a corrosion inhibitor. It is utilized to remove hydrogen sulfide and carbon dioxide from natural gas. Further, it is an intermediate used in the rubber chemicals, as a humectant and a softening agent, and as an emulsifier and dispersing agent in agricultural chemicals. In addition to this, it is used in cutting oils, cleaners, soaps, polishers, and pharmaceuticals.

Solubility
Miscible with water.

Notes
Hygroscopic and air sensitive. Incompatible with strong oxidizing agents, strong acids, copper, zinc, carbon dioxide and iron.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text