N,N'-Diacetylhydrazine, 98%, Thermo Scientific Chemicals
N,N'-Diacetylhydrazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Diacetylhydrazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A13393.18
also known as A13393-18
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
N,N'-Diacetylhydrazine, 98%, Thermo Scientific Chemicals
Catalog numberA13393.18
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS3148-73-0
IUPAC NameN'-acetylacetohydrazide
Molecular FormulaC4H8N2O2
InChI KeyZLHNYIHIHQEHJQ-UHFFFAOYSA-N
SMILESCC(=O)NNC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)134.0-143.0?C
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only