Thermo Scientific Chemicals

3,4-Dimethoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A13396.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A13396.14
25 g, Each
Quantity
Catalog number: A13396.14
also known as A13396-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-03-8
IUPAC Name
(3,4-dimethoxyphenyl)methanol
Molecular Formula
C9H12O3
InChI Key
OEGPRYNGFWGMMV-UHFFFAOYSA-N
SMILES
COC1=CC=C(CO)C=C1OC
Form
Liquid
Refractive Index
1.5505-1.5555 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow to orange

Description

3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

Solubility
Fully miscible with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text