Thermo Scientific Chemicals

Thiosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13401.18
50 g, Each
Thermo Scientific Chemicals

Thiosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13401.18
50 g, Each
Quantity
Catalog number: A13401.18
also known as A13401-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
147-93-3
IUPAC Name
2-sulfanylbenzoic acid
Molecular Formula
C7H6O2S
InChI Key
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1S
Appearance (Color)
White to cream to yellow
Assay (Argentometric Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
163.0-169.0?C
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.

Solubility
Soluble in dimethyl sulfoxide, alcohol and ether. Slightly soluble in water, ethanol and alkanes.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text