Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
Solubility
Soluble in dimethyl sulfoxide, alcohol and ether. Slightly soluble in water, ethanol and alkanes.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Kork, S.; Yilmaz, G.; Yagci, Y. Poly(vinyl alcohol)-Thioxanthone as One-Component Type II Photoinitiator for Free Radical Polymerization in Organic and Aqueous Media. Macromol. Rapid Commun. 2015, 36 (10), 923-928.
Lin, X.; Xu, S.; Wang, C.; Wang, Z.; Cui, Y. Synthesis of thiosalicylic acid-capped CdTe quantum dots. RSC Adv. 2014, 4 (10), 4993-4997.