Thermo Scientific Chemicals

2-Phenylethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A13402.AD
50 mL, Each
Thermo Scientific Chemicals

2-Phenylethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A13402.AD
50 mL, Each
Quantity
Catalog number: A13402.AD
also known as A13402-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64-04-0
IUPAC Name
2-phenylethan-1-amine
Molecular Formula
C8H11N
InChI Key
BHHGXPLMPWCGHP-UHFFFAOYSA-N
SMILES
NCCC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5320-1.5350 @ 20°C

Description

Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethylamine is used in manufacturing anti-depression agents and anti diabetic drugs. It is also used as the goodds for drug jiangtangling intermediate, also used for other organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Store under inert gas. Keep away from acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide (CO2).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text