Thermo Scientific Chemicals

2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals

Catalog number: A13406.18
50 g, Each
Thermo Scientific Chemicals

2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals

Catalog number: A13406.18
50 g, Each
Quantity
Catalog number: A13406.18
also known as A13406-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3141-27-3
IUPAC Name
2,5-dibromothiophene
Molecular Formula
C4H2Br2S
InChI Key
KBVDUUXRXJTAJC-UHFFFAOYSA-N
SMILES
BrC1=CC=C(Br)S1
Appearance (Color)
Clear pale yellow to yellow to brown
Form
Liquid
Assay from Suppliers CofA
≥94.0%

Description

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text