2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.
Solubility
It is soluble in chloroform and methanol. It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
RUO – Research Use Only
General References:
- JWP Lin.; LP Dudek. Synthesis and properties of poly (2, 5-thienylene). Journal of Polymer Science. 1980, 18,(9), 2869-2873.
- . J Nakayama.; N Katano.; Y Shimura. Preparation of Oligo (thio-2, 5-thienylenes). J. Org. Chem. 1996, 61,(21), 7608-7610.
- For selective Mg exchange at the 2-position, see: J. Org. Chem., 65, 4618 (2000).