5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water, Thermo Scientific Chemicals
5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A13410.14
also known as A13410-14
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
5-Azacytosine, 98% (dry wt.), may cont. up to ca 7% water, Thermo Scientific Chemicals
Catalog numberA13410.14
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS931-86-2
IUPAC Name6-amino-1,2-dihydro-1,3,5-triazin-2-one
Molecular FormulaC3H4N4O
InChI KeyMFEFTTYGMZOIKO-UHFFFAOYSA-N
SMILESNC1=NC=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (dry wt. basis)
Water Content (Karl Fischer Titration)≤7%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only