Thermo Scientific Chemicals

2-Bromo-4'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13415.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-4'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A13415.14
25 g, Each
Quantity
Catalog number: A13415.14
also known as A13415-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
2632-13-5
IUPAC Name
2-bromo-1-(4-methoxyphenyl)ethan-1-one
Molecular Formula
C9H9BrO2
InChI Key
XQJAHBHCLXUGEP-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(=O)CBr
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to pale brown
Melting Point (clear melt)
67-73?C

Description

2-Bromo-4'-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.

Solubility
It is soluble in DMSO, water (partly miscible), most organic solvents, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician