Thermo Scientific Chemicals

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals

Catalog number: A13418.06
5 g, Each
Thermo Scientific Chemicals

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, 97%, Thermo Scientific Chemicals

Catalog number: A13418.06
5 g, Each
Quantity
Catalog number: A13418.06
also known as A13418-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3162-96-7
IUPAC Name
6-methoxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-7,8-diol
Molecular Formula
C14H18O6
InChI Key
VVSWDMJYIDBTMV-UHFFFAOYNA-N
SMILES
COC1OC2COC(OC2C(O)C1O)C1=CC=CC=C1
Appearance (Color)
White
Melting Point (clear melt)
161.0-170.0?C
Optical Rotation
110.0 ± 0.5? (C=2 in chloroform)
Form
Powder or fused solid
Assay (GC)
≥96.0%

Description

Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4,6-O-benzylidene-alpha-D-glucopyranoside, is used as a chiral building block and important intermediate in the preparation of different sugars. It is also used as an intermediate in chemical, pharmaceutical intermediate, pharmaceutical research and reagent.

Solubility
It is soluble in water, acetone, benzene, ethyl ether

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text