Thermo Scientific Chemicals

trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer, Thermo Scientific Chemicals

Catalog number: A13419.14
25 g, Each
Thermo Scientific Chemicals

trans,trans-2,4-Undecadienal, 90+%, remainder mainly trans,cis isomer, Thermo Scientific Chemicals

Catalog number: A13419.14
25 g, Each
Quantity
Catalog number: A13419.14
also known as A13419-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30361-29-6
IUPAC Name
(2E,4E)-undeca-2,4-dienal
Molecular Formula
C11H18O
InChI Key
UVIUIIFPIWRILL-XBLVEGMJSA-N
SMILES
CCCCCCC=CC=CC=O
Form
Liquid
Refractive Index
1.5080-1.5160 @ 20?C
Assay (GC)
>90.0% (major isomer)
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text