Thermo Scientific Chemicals

Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals

Catalog number: A13429.14
25 g, Each
Thermo Scientific Chemicals

Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals

Catalog number: A13429.14
25 g, Each
Quantity
Catalog number: A13429.14
also known as A13429-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
621-79-4
IUPAC Name
(2E)-3-phenylprop-2-enamide
Molecular Formula
C9H9NO
InChI Key
APEJMQOBVMLION-VOTSOKGWSA-N
SMILES
NC(=O)\C=C\C1=CC=CC=C1
Form
Crystalline powder
Appearance (Color)
White to cream
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Melting Point
144.0-150.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text