Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals
Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A13429.22
also known as A13429-22
Price (USD)/ Each
403.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
403.00
Add to cart
Cinnamamide, predominantly trans, 97%, Thermo Scientific Chemicals
Catalog numberA13429.22
Price (USD)/ Each
403.00
-
Add to cart
Chemical Identifiers
CAS621-79-4
IUPAC Name(2E)-3-phenylprop-2-enamide
Molecular FormulaC9H9NO
InChI KeyAPEJMQOBVMLION-VOTSOKGWSA-N
SMILESNC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White to cream
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Melting Point144.0-150.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only