1,2-Dichloro-4-iodobenzene, 98%, Thermo Scientific Chemicals
1,2-Dichloro-4-iodobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Dichloro-4-iodobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13440.09
also known as A13440-09
Price (USD)/ Each
37.90
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
37.90
Add to cart
1,2-Dichloro-4-iodobenzene, 98%, Thermo Scientific Chemicals
Catalog numberA13440.09
Price (USD)/ Each
37.90
-
Add to cart
Chemical Identifiers
CAS20555-91-3
IUPAC Name1,2-dichloro-4-iodobenzene
Molecular FormulaC6H3Cl2I
InChI KeyNADPFZNWCQIJJW-UHFFFAOYSA-N
SMILESClC1=CC=C(I)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormFused solid
Melting Point (clear melt)27.0-34.0°C
Appearance (Color)Pale yellow to pale browm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only