Thermo Scientific Chemicals

Tetramethyl methylenediphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13441.22
100 g, Each
Thermo Scientific Chemicals

Tetramethyl methylenediphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13441.22
100 g, Each
Quantity
Catalog number: A13441.22
also known as A13441-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16001-93-7
IUPAC Name
dimethyl [(dimethoxyphosphoryl)methyl]phosphonate
Molecular Formula
C5H14O6P2
InChI Key
XAVFZUKFLWOSOS-UHFFFAOYSA-N
SMILES
COP(=O)(CP(=O)(OC)OC)OC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid or viscous liquid
Assay (GC)
≥98.0%
Refractive Index
1.4525-1.4565 @ 20?C

Description

Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text