It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Antonio Trincone; Edoardo Pagnotta. Efficient chemoselective synthesis of 3,4'-dihydroxypropiophenone 3-O-β-D-glucoside by thermophilic β-glycosidase from Sulfolobus solfataricus. Biotechnology Letters. 1995, 17 (1), 45-48.
- Elwin E. Harris; James D'Ianni; Homer Adkins. Reaction of Hardwood Lignin with Hydrogen. J. Am. Chem. Soc, 1938, 60 (6), 1467-1470.