Thermo Scientific Chemicals

Sodium octanoate, 96%, Thermo Scientific Chemicals

Catalog number: A13451.22
100 g, Each
Thermo Scientific Chemicals

Sodium octanoate, 96%, Thermo Scientific Chemicals

Catalog number: A13451.22
100 g, Each
Quantity
Catalog number: A13451.22
also known as A13451-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1984-06-1
IUPAC Name
sodium octanoate
Molecular Formula
C8H15NaO2
InChI Key
BYKRNSHANADUFY-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCC([O-])=O
Form
Powder or crystals or crystalline powder
Identification (FTIR)
Conforms (non-U.S. specification)
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)
≤3%

Description

Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text