Thermo Scientific Chemicals

1,3,5-Triisopropylbenzene, 95%, Thermo Scientific Chemicals

Catalog number: A13453.30
250 g, Each
Thermo Scientific Chemicals

1,3,5-Triisopropylbenzene, 95%, Thermo Scientific Chemicals

Catalog number: A13453.30
250 g, Each
Quantity
Catalog number: A13453.30
also known as A13453-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
717-74-8
IUPAC Name
1,3,5-tris(propan-2-yl)benzene
Molecular Formula
C15H24
InChI Key
VUMCUSHVMYIRMB-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C
Assay (GC)
≥94.0%
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4855-1.4915 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text