Thermo Scientific™

4-Iodophenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13455.06
5 g, Each
Thermo Scientific™

4-Iodophenol, 98+%, Thermo Scientific Chemicals

Catalog number: A13455.06
5 g, Each
Quantity
Catalog number: A13455.06
also known as A13455-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
540-38-5
IUPAC Name
4-iodophenol
Molecular Formula
C6H5IO
InChI Key
VSMDINRNYYEDRN-UHFFFAOYSA-N
SMILES
OC1=CC=C(I)C=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
90.0-96.0?C
Appearance (Color)
White to pale cream to cream to brown
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, and polarizing films for Liquid Crystal Display (LCD) chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display (LCD) chemicals.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text