Thermo Scientific Chemicals

5-Fluorouracil, 99%, Thermo Scientific Chemicals

Catalog number: A13456.14
25 g, Each
Thermo Scientific Chemicals

5-Fluorouracil, 99%, Thermo Scientific Chemicals

Catalog number: A13456.14
25 g, Each
Quantity
Catalog number: A13456.14
also known as A13456-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-21-8
IUPAC Name
5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular Formula
C4H3FN2O2
InChI Key
GHASVSINZRGABV-UHFFFAOYSA-N
SMILES
FC1=CNC(=O)NC1=O
Form
Crystalline powder
Appearance (Color)
White to pale cream
Assay from Suppliers CofA
≥98.5%

Description

5-Fluorouracil is used as an antitumor agent in the treatment of anal, breast, colorectal, oesophageal, stomach, pancreatic and skin cancers. It finds application as a suicide inhibitor due to its irreversible inhibition of thymidylate synthase. It is also used in the treatment of actinic keratoses and bowen's disease. Further, it serves as a potent antineoplastic agent in clinical use. In addition to this, it acts as a DNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide, methanol, hot water, ammonium hydroxide, sodium hydroxide, ethanol and water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text