Thermo Scientific Chemicals

Methyl 4-(methylamino)benzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13460.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-(methylamino)benzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13460.14
25 g, Each
Quantity
Catalog number: A13460.14
also known as A13460-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18358-63-9
IUPAC Name
methyl 4-(methylamino)benzoate
Molecular Formula
C9H11NO2
InChI Key
LLAMGYUWYUMHCH-UHFFFAOYSA-N
SMILES
CNC1=CC=C(C=C1)C(=O)OC
Appearance (Color)
White to cream to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
92-98?C (≤2.5?C range)
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
It is slightly soluble in water. (1.2 g/L) (25°C insoluble in hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text