Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13461.30
250 g, Each
Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13461.30
250 g, Each
Quantity
Catalog number: A13461.30
also known as A13461-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5965-83-3
IUPAC Name
2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular Formula
C9H14O6S
InChI Key
NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
14-22%
Identification (FTIR)
Conforms
Melting Point (clear melt)
99-112?C

Description

5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([?]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Sulfosalicylic acid dihydrate may be employed for the quantification of doxorubicin derived from PEGylated liposomal doxorubicin (Doxil) and its major metabolite in human plasma by HPLC. It may be used in the preparation of poly[[tris(di-2-pyridylamine)tris([μ]-5-sulfonatosalicylato)tricopper(II)] trihydrate]. Also used as an intermediate synthesis of bulk drugs, surfactant and additive for lubricating grease.

Solubility
Soluble in water

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from strong bases and oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text