Thermo Scientific Chemicals

Sulfolane, 99%, Thermo Scientific Chemicals

Catalog number: A13466.0I
5000 g, Each
Thermo Scientific Chemicals

Sulfolane, 99%, Thermo Scientific Chemicals

Catalog number: A13466.0I
5000 g, Each
Quantity
Catalog number: A13466.0I
also known as A13466-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
126-33-0
IUPAC Name
1λ⁶-thiolane-1,1-dione
Molecular Formula
C4H8O2S
InChI Key
HXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILES
O=S1(=O)CCCC1
Refractive Index
1.4805-1.4850 @ melt
Form
Liquid as melt
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.5%
Appearance (Color)
Clear colorless to yellow

Description

Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.

Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text