Thermo Scientific Chemicals

Di-n-octyl phenylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: A13468.06
5 g, Each
Thermo Scientific Chemicals

Di-n-octyl phenylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: A13468.06
5 g, Each
Quantity
Catalog number: A13468.06
also known as A13468-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1754-47-8
IUPAC Name
dioctyl phenylphosphonate
Molecular Formula
C22H39O3P
InChI Key
HAKMAMKAFTZXOZ-UHFFFAOYSA-N
SMILES
CCCCCCCCOP(=O)(OCCCCCCCC)C1=CC=CC=C1
Refractive Index
1.4760-1.4810 @ 20°C
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%

Description

Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-octyl phenylphosphonate acts as a plasticizer, which is useful in ion-selective electrodes.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text