Thermo Scientific Chemicals

Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: A13470.0B
1000 g, Each
Thermo Scientific Chemicals

Ethyl 4-bromobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: A13470.0B
1000 g, Each
Quantity
Catalog number: A13470.0B
also known as A13470-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2969-81-5
IUPAC Name
ethyl 4-bromobutanoate
Molecular Formula
C6H11BrO2
InChI Key
XBPOBCXHALHJFP-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCBr
Identification (FTIR)
Conforms
Refractive Index
1.4540-1.4580 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobutyrate is used as a building block for the preparation of firefly luciferase inhibitor-conjucated peptide derivatives. It finds application in the synthesis of poly(2,6,-dimethyl-1,4-phehylene oxide) and thieno(2,3-b)-azepin-4-ones.

Solubility
Immiscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text