Thermo Scientific Chemicals

1,4-Diphenylbutadiyne, 99%, Thermo Scientific Chemicals

Catalog number: A13471.22
100 g, Each
Thermo Scientific Chemicals

1,4-Diphenylbutadiyne, 99%, Thermo Scientific Chemicals

Catalog number: A13471.22
100 g, Each
Quantity
Catalog number: A13471.22
also known as A13471-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
886-66-8
IUPAC Name
(4-phenylbuta-1,3-diyn-1-yl)benzene
Molecular Formula
C16H10
InChI Key
HMQFJYLWNWIYKQ-UHFFFAOYSA-N
SMILES
C1=CC=C(C=C1)C#CC#CC1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
84.0-91.0°C

Description

1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

Solubility
Sparingly soluble in water(5.5E-4 g/L) (25°C). Soluble in toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text