Thermo Scientific™

Xanthydrol, 98+%, Thermo Scientific Chemicals

Catalog number: A13476.06
5 g, Each
Thermo Scientific™

Xanthydrol, 98+%, Thermo Scientific Chemicals

Catalog number: A13476.06
5 g, Each
Quantity
Catalog number: A13476.06
also known as A13476-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-46-0
IUPAC Name
9H-xanthen-9-ol
Molecular Formula
C13H10O2
InChI Key
JFRMYMMIJXLMBB-UHFFFAOYSA-N
SMILES
OC1C2=CC=CC=C2OC2=CC=CC=C12
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.0%

Description

Useful in tests for urea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text