Thermo Scientific Chemicals

cis-Cyclooctene, 95%, stab., Thermo Scientific Chemicals

Catalog number: A13477.AP
500 mL, Each
Thermo Scientific Chemicals

cis-Cyclooctene, 95%, stab., Thermo Scientific Chemicals

Catalog number: A13477.AP
500 mL, Each
Quantity
Catalog number: A13477.AP
also known as A13477-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
931-87-3
IUPAC Name
(Z)-cyclooctene
Molecular Formula
C8H14
InChI Key
URYYVOIYTNXXBN-UPHRSURJSA-N
SMILES
C1CCCC=C/CC1
Form
Liquid
Comment
Stabilized with 100-200 ppm Irganox® 1076 FD
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Refractive Index
1.4660-1.4720 @20?C

Description

cis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text