Thermo Scientific Chemicals

Iron(II) oxalate dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A13479.30
250 g, Each
Thermo Scientific Chemicals

Iron(II) oxalate dihydrate, 99%, Thermo Scientific Chemicals

Catalog number: A13479.30
250 g, Each
Quantity
Catalog number: A13479.30
also known as A13479-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6047-25-2
IUPAC Name
λ²-iron(2+) dihydrate oxalate
Molecular Formula
C2H4FeO6
InChI Key
NPLZZSLZTJVZSX-UHFFFAOYSA-L
SMILES
O.O.[Fe++].[O-]C(=O)C([O-])=O
Appearance (Color)
Yellow to brown
Assay (Redox Titration)
≥98.5 to ≤101.5%
Form
Powder
Identification (FTIR)
Conforms

Description

It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store at room temperature. It is not intended for diagnostic and therapeutic purposes. Avoid contamination with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text