Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Chiricof, R.D.; Cowell, A.B.; Good, W.D.; Klots, T.D.; Knipmeyer, S.E.; Nguyen, A.; Rau, A.P.; Reynolds, J.W.; Smith, N.K.; Steele, W.V. Heat capacities, enthalpy increments, phase transitions, and derived thermodynamic functions for the condensed phases of bicyclohexyl between the temperatures 6 K and 440 K ☆. The Journal of Chemical Thermodynamics. 1998, 30, (12), 1423-1439.
- Eva Garcia Fidalgo.; Laurent Plasseraud.; Georg SUss-Fink. Catalysis in aqueous solution: Hydrogenation of benzene derivatives catalyzed by (η6-C6H6)2Ru2Cl4. Journal of Molecular Catalysis A: Chemical. 2001, 132, (1), 5-12.