Thermo Scientific Chemicals

Bicyclohexyl, 98%, Thermo Scientific Chemicals

Catalog number: A13481.18
50 g, Each
Thermo Scientific Chemicals

Bicyclohexyl, 98%, Thermo Scientific Chemicals

Catalog number: A13481.18
50 g, Each
Quantity
Catalog number: A13481.18
also known as A13481-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-51-3
IUPAC Name
1,1'-bi(cyclohexane)
Molecular Formula
C12H22
InChI Key
WVIIMZNLDWSIRH-UHFFFAOYSA-N
SMILES
C1CCC(CC1)C1CCCCC1
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5% (U.K. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Identification (FTIR)
Conforms (U.K. specification)
Proton NMR
Conforms to structure (U.S. specification)

Description

Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text