Thermo Scientific Chemicals

trans-Anethole, 98+%, Thermo Scientific Chemicals

Catalog number: A13482.14
25 g, Each
Thermo Scientific Chemicals

trans-Anethole, 98+%, Thermo Scientific Chemicals

Catalog number: A13482.14
25 g, Each
Quantity
Catalog number: A13482.14
also known as A13482-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4180-23-8
IUPAC Name
1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene
Molecular Formula
C10H12O
InChI Key
RUVINXPYWBROJD-ONEGZZNKSA-N
SMILES
COC1=CC=C(\C=C\C)C=C1
Comment
Residual toluene <100ppm
Refractive Index
1.5580-1.5625 as melt
Appearance (Color)
Translucent/colourless to pale yellow
Form
Low-melting solid or clear liquid as melt
Assay (GC)
≥98.0%

Description

trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.

Solubility
Soluble in benzene, ethyl acetate, acetone, carbon disulfide.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text