Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13488.0B
1000 g, Each
Thermo Scientific Chemicals

2-Chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13488.0B
1000 g, Each
Quantity
Catalog number: A13488.0B
also known as A13488-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89-98-5
IUPAC Name
2-chlorobenzaldehyde
Molecular Formula
C7H5ClO
InChI Key
FPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1C=O
Free acid (titration)
≤2.25%
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.5640-1.5690 @ 20?C
Form
Liquid

Description

2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

Solubility
Soluble in water (1.8g/L), alcohol, ether, benzene, and acetone.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text