Thermo Scientific Chemicals

1-Chloro-3-iodopropane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A13489.14
25 g, Each
Thermo Scientific Chemicals

1-Chloro-3-iodopropane, 98%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A13489.14
25 g, Each
Quantity
Catalog number: A13489.14
also known as A13489-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6940-76-7
IUPAC Name
1-chloro-3-iodopropane
Molecular Formula
C3H6ClI
InChI Key
SFOYQZYQTQDRIY-UHFFFAOYSA-N
SMILES
ClCCCI
Appearance (Color)
Clear, colourless to yellow to orange or pale brown
Assay (GC)
≥97.5%
Stabilizer
Copper powder
Form
Liquid
Refractive Index
1.5440-1.5490 @ 20?C

Description

1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text