Thermo Scientific Chemicals

2,3-Dihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13490.14
25 g, Each
Thermo Scientific Chemicals

2,3-Dihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13490.14
25 g, Each
Quantity
Catalog number: A13490.14
also known as A13490-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24677-78-9
IUPAC Name
2,3-dihydroxybenzaldehyde
Molecular Formula
C7H6O3
InChI Key
IXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILES
OC1=CC=CC(C=O)=C1O
Appearance (Color)
Pale yellow to yellow-green or cream to brown or grey
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Clarity
5% w/v in methanol: clear solution
Melting Point (clear melt)
103-109?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text