Thermo Scientific Chemicals

2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals

Catalog number: A13496.14
25 g, Each
Thermo Scientific Chemicals

2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals

Catalog number: A13496.14
25 g, Each
Quantity
Catalog number: A13496.14
also known as A13496-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2295-31-0
IUPAC Name
1,3-thiazolidine-2,4-dione
Molecular Formula
C3H3NO2S
InChI Key
ZOBPZXTWZATXDG-UHFFFAOYSA-N
SMILES
O=C1CSC(=O)N1
Melting Point (clear melt)
121-130?C
Form
Crystalline powder or crystals
Assay (HPLC)
≥98.5%
Appearance (Color)
White to cream or pale yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

Solubility
Soluble in water (30 mg/ml at 3°C), methanol, DMSO, ethanol, and ethyl ether.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text