Thermo Scientific Chemicals

(1-Hexadecyl)pyridinium chloride monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13499.22
100 g, Each
Thermo Scientific Chemicals

(1-Hexadecyl)pyridinium chloride monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13499.22
100 g, Each
Quantity
Catalog number: A13499.22
also known as A13499-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6004-24-6
IUPAC Name
1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
Molecular Formula
C21H40ClNO
InChI Key
NFCRBQADEGXVDL-UHFFFAOYSA-M
SMILES
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Melting Point (clear melt)
81.0-87.0?C

Description

(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexadecyl)pyridinium chloride monohydrate is used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water. And it is used for detergent-based extraction of membrane-associated enzymes such as alkaline phosphatase, and in extraction of chromium and palladium.

Solubility
Soluble in water, alcohol, chloroform

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text