Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.
Solubility
Soluble in water and ethanol. Insoluble in ether and chloroform.
Notes
Incompatible with strong oxidizing agents, acids, bases and strong reducing agents.
RUO – Research Use Only
General References:
- With the cycloalkenylation agent (1-Ethoxycarbonyl cyclopropyl) triphenyl phosphonium tetrafluoroborate, A18305, gives a bridgehead lactam (pyrrolizinone) which is an intermediate in a diastereoselective synthesis of (±)-isoretronecanol: Liebigs Ann. Chem., 521 (1983):
- Peng, H.; Kather, M.; Rübsam, K.; Jakob, F.; Schwaneberg, U.; Pich, A. Water-Soluble Reactive Copolymers Based on Cyclic N-Vinylamides with Succinimide Side Groups for Bioconjugation with Proteins. Macromolecules 2015, 48 (13), 4256-4268.
- Habibi, A.; Soltanloo, Z.; Beiraghi, A.; Valizadeh, Y. One-pot synthesis of new dye derivatives of succinimide via three-component reaction of alkylidene Meldrum's acid, isocyanides and p-aminoazobenzene derivatives. Dyes Pigm. 2015, 113, 481-486.