Thermo Scientific Chemicals

Succinimide, 98+%, Thermo Scientific Chemicals

Catalog number: A13503.0I
5000 g, Each
Thermo Scientific Chemicals

Succinimide, 98+%, Thermo Scientific Chemicals

Catalog number: A13503.0I
5000 g, Each
Quantity
Catalog number: A13503.0I
also known as A13503-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-56-8
IUPAC Name
pyrrolidine-2,5-dione
Molecular Formula
C4H5NO2
InChI Key
KZNICNPSHKQLFF-UHFFFAOYSA-N
SMILES
O=C1CCC(=O)N1
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
119-127?C
Form
Crystalline powder or flakes

Description

Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

Solubility
Soluble in water and ethanol. Insoluble in ether and chloroform.

Notes
Incompatible with strong oxidizing agents, acids, bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text