Thermo Scientific Chemicals

Piperine, 98%, Thermo Scientific Chemicals

Catalog number: A13510.06
5 g, Each
Thermo Scientific Chemicals

Piperine, 98%, Thermo Scientific Chemicals

Catalog number: A13510.06
5 g, Each
Quantity
Catalog number: A13510.06
also known as A13510-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-62-2
IUPAC Name
(2E,4E)-5-(2H-1,3-benzodioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one
Molecular Formula
C17H19NO3
InChI Key
MXXWOMGUGJBKIW-YPCIICBESA-N
SMILES
O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1
Appearance (Color)
White to pale cream to pale yellow
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
128.0-134.0?C

Description

Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

Solubility
Soluble in water(0.04g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text