L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
Thermo Scientific Chemicals

L-Aspartic acid, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
Catalog number A13520.0I
also known as A13520-0I
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
Add to cart
L-Aspartic acid, 98+%
Catalog numberA13520.0I
Price (USD)/ Each
309.65
Online exclusive
344.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS56-84-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0% (dry wt. basis)
Loss on Drying≤0.5% (105°C)
Identification (FTIR)Conforms
View more
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

Solubility
Soluble in water(4.5g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Flaig, R.; Koritsanszky, T.; Zobel, D.; Luger, P. Topological Analysis of the Experimental Electron Densities of Amino Acids. 1. d,l-Aspartic Acid at 20 K. J. Am. Chem. Soc. 1998, 120, (10), 2227-2238.
  2. Xiao-Lin Wang.; Ai-Ling Ying.; Wei-Ning Wang. Nanofiltration of l-phenylalanine and l-aspartic acid aqueous solutions. Journal of Membrane Science. 2002, 196, (1), 59-67.
  3. Readily forms ß-lactam derivatives whichare valuable intermediates for the synthesis of a variety of products, including thienamycin: J. Am. Chem. Soc., 102, 6163 (1980); Tetrahedron Lett., 23, 2293 (1982).