Thermo Scientific Chemicals

2-Aminobenzothiazole, 98%, Thermo Scientific Chemicals

Catalog number: A13521.36
500 g, Each
Thermo Scientific Chemicals

2-Aminobenzothiazole, 98%, Thermo Scientific Chemicals

Catalog number: A13521.36
500 g, Each
Quantity
Catalog number: A13521.36
also known as A13521-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
136-95-8
IUPAC Name
1,3-benzothiazol-2-amine
Molecular Formula
C7H6N2S
InChI Key
UHGULLIUJBCTEF-UHFFFAOYSA-N
SMILES
NC1=NC2=CC=CC=C2S1
Form
Powder
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text